Learn More
Invitrogen™ eBioscience™ Streptavidin PE-Cyanine7 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
Brand: Invitrogen™ 25-4317-82
Alert:
Contains Sodium azide
Description
Specifications
0.2 mg/mL | |
100 μg | |
eBioscience™ | |
Biotin |
PE-Cy7 | |
Liquid | |
Streptavidin Conjugate (Fluorescent) | |
PE-Cy7 |
For Research Use Only