Learn More
Cole-Parmer™ Drum for DR-200 Series Rotating Shaker System Drives
Click to view available options
No. of Positions:
1
2
4
240
For Use With (Equipment):
1.5 mL Microcentrifuge Tubes
250 mL Flasks or Bottles
Bottles
Clip Plates
One Bottle, Figure-8 Motion
Unit Size:
Each
Description
- Change your rotator's capacity with interchangeable drums
- Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Specifications
| Product Type | Drum Test Tube Holder |
| For Use With (Application) | For changing your rotator's capacity |
| For Use With (Equipment) | 250 mL Flasks or Bottles |
| No. of Positions | 2 |