missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinic acid monomethyl ester, ≥95%, MP Biomedicals™
Click to view available options
Quantity:
10 g
Specifications
Specifications
CAS | 3878-55-5 |
Color | White |
Molecular Formula | C5H8O4 |
Quantity | 10 g |
Synonym | monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester |
InChI Key | JDRMYOQETPMYQX-UHFFFAOYSA-N |
SMILES | COC(=O)CCC(=O)O |
IUPAC Name | 4-methoxy-4-oxobutanoic acid |
Molecular Weight (g/mol) | 132.115 |
PubChem CID | 77487 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'