missing translation for 'onlineSavingsMsg'
Learn More

Succinic Anhydride, >99%, MP Biomedicals™

Product Code. 11483331 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 108-30-5
Color White
Density 1.572g/cm3
Quantity 1 kg
Physical Form Solid
Assay Percent Range >99%
Molecular Formula C4H4O3
MDL Number MFCD00005525
Synonym succinic anhydride, dihydrofuran-2,5-dione, succinic acid anhydride, succinyl oxide, 2,5-diketotetrahydrofuran, dihydro-2,5-furandione, butanedioic anhydride, 2,5-furandione, dihydro, succinyl anhydride, rikacid sa
InChI Key RINCXYDBBGOEEQ-UHFFFAOYSA-N
SMILES O=C1CCC(=O)O1
IUPAC Name oxolane-2,5-dione
Molecular Weight (g/mol) 100.07
PubChem CID 7922
ChEBI CHEBI:36595
Formula Weight 100.1
Chemical Name or Material Succinic Anhydride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Succinic Anhydride, >99%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.