Learn More
Heidolph Instruments Test Tube Holding Device
For use with Reax top and Reax control
Brand: Heidolph Instruments 549-20000-00
This item is not returnable.
View return policy
Description
Attachment to hold one test tube safely for continuous shakingSpecifications
115/230 V | |
Supporting Rod | |
For Test Tube |