Learn More
Eppendorf™ Swing Out Rotor for Benchtop Centrifuge
Shop All Eppendorf Products
missing translation for 'orderingAttributeHoverText'
Capacity:
0.2 to 500 mL
5 to 25 mL
For Use With (Equipment):
5702/5702 R/5702 RH Centrifuge
5810/5810R Centrifuge
missing translation for 'unitSize'
Each
Description
- For use with Eppendorf Centrifuges 5702
- 5702 family of low speed, general purpose centrifuges was especially designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs. They are compact in design so that one can fit comfortably on every lab bench, quiet in operation to benefit your work environment.
Specifications
Specifications
| Type | Swing-out Rotor without Basket |
| Capacity | 5 to 25 mL |
| Max. RCF | 3,000 x g |
| Max. Speed | 4,400 rpm |
| For Use With (Equipment) | 5702/5702 R/5702 RH Centrifuge |
| No. of Places | 15 |