Learn More
BINDER™ Swivel Casters
Product Code. 30238233 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 260, KB 470, KB 720, KB 1060
KB PRO Series
KBF PRO Series
KBF Series
Unit Size:
Each
Product Code. 30238233
Brand: BINDER™ 80122543
Only null left
Add to basket
Add to basket
Height-adjustable swivel casters
Height-adjustable swivel casters for horizontal alignment of unit on uneven floors.
Specifications
For Use With (Equipment) KBF Series
Material Steel
Type Castor Frame