missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptophysin Monoclonal Antibody (SYP02), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511575
This item is not returnable.
View return policy
Description
MA5-11575 targets Synaptophysin in IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-11575 immunogen is a synthetic peptide encoding a region near the C-terminal end of the synaptophysin protein.
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.Specifications
Synaptophysin | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
P07825, P08247, Q62277 | |
Syp | |
A synthetic peptide encoding a region near the C-terminal end of the synaptophysin protein | |
RUO | |
20977, 24804, 6855 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
SYP02 | |
Unconjugated | |
Syp | |
A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1 | |
Mouse | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |