Learn More
Syngene™ Long Wave UV Tube
GREENER_CHOICE
Syngene™ Long Wave UV Tube for use in UV Transilluminators.
Brand:  Syngene™ LWT8
Product Code. 12883998
  • 27.27 EUR / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Description
  • For use with all GX and GE model transilluminators
  • 365nm wavelength
  • Long wave
Specifications
Specifications
365 nm
All GX/GE model transilluminators
Longwave UV Tube