Learn More
Sartorius Tacta™ Mechanical Pipettes
Shop All Sartorius Products
Click to view available options
Number of Channels:
1
8
12
Volume (Metric):
0.1 to 3 μL
0.5 to 10 μL
10 to 100 μL
100 to 1000 μL
1000 to 10,000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 100 μL
500 to 5000 μL
Color:
Blue
Gray
Green
Orange
Red
Yellow
Unit Size:
Each
Description
- Comfortable to hold due to the ergonomically designed handle
- Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
- Controlled and smooth tip ejection with the new Sartorius Optiject technology
- Calibration adjustment to provide accurate results for various liquid types
- Easy to clean, with only three parts to disassemble
- Reliable results, even over lengthy pipetting periods
Specifications
Specifications
| Compatible Tips | Optifit 200 μL, 350 μL, SafetySpace Filter Tips 120 μL |
| Adjustable Tip Spacing | Yes |
| Autoclavable | Autoclavable |
| Color | Yellow |
| Disposable | Reusable |
| Grip Type | Ergonomic |
| Material | Polypropylene |
| Number of Channels | 12 |
| Product Line | Tacta |
| Programmability | No |
| Show More |