Learn More
Thermo Scientific™ Tango™ PTAFR-bla U2OS Cells
The Tango™ PTAFR-bla U2OS cells contain the human Platelet Activating Factor Receptor (PTAFR) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand:  Thermo Scientific™ K1789
Product Code. 10698324
  • 16130.00 EUR / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Description
The Tango™ PTAFR-bla U2OS cells contain the human Platelet Activating Factor Receptor (PTAFR) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ PTAFR-bla U2OS cells are functionally validated for Z' and EC50 concentrations of β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine. In addition, Tango™ PTAFR-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_000952
Dividing Cells
Native Tail
FRET
PTAFR-bla U2OS cells, PTAFR-bla U2OS Assay Protocol, Certificate of Analysis
GeneBLAzer™
End Point
PTAFR
U2OS
GPCR
Cell-based beta-arrestin recruitment
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• Tango™ PTAFR-bla U2OS cells
• Tango™ PTAFR-bla U2OS Assay Protocol
• Certificate of Analysis
The Tango™ PTAFR-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Fluorescence
PTAFR
Pool of Stable Cells
8 x 106 Cells
BLA (Beta-Lactamase)
Dividing Cells
Tango™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.