Learn More
TCR V beta 8b Monoclonal Antibody (MX-6), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen TCR2750
Additional Details : Weight : 0.05000kg Transport : UN number : 3276 Chem class : 6.1 Pack group : III
Description
TCR2750 targets TCR V beta 8b in FACS, IHC(F), IP, and TCA applications and shows reactivity with Human samples. The TCR2750 immunogen is human TCR Vbeta8.
The ability of T cell receptors (TCR) to discriminate foreign from self-peptides presented by major histocompatibility complex (MHC) class II molecules is essential for an effective adaptive immune response. TCR recognition of self-peptides has been linked to autoimmune disease. Mutant self-peptides have been associated with tumors. Engagement of TCRs by a family of bacterial toxins know as superantigens has been responsible for toxic shock syndrome. Autoantibodies to V beta segments of T cell receptors have been isolated from patients with rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE). The autoantibodies block TH1-mediated inflammatory autodestructive reactions and are believed to be a method by which the immune system compensates for disease (ref5). T Cell and TCR Diversity Most human T cells express the TCR alpha-beta and either CD4 or CD8 molecule (single positive, SP). A small number of T cells lack both CD4 and CD8 (double negative, DN). Increased percentages of alpha-beta DN T cells have been identified in some autoimmune and immunodeficiency disorders. Gamma-delta T cells are primarily found within the epithelium. They show less TCR diversity and recognize antigens differently than alpha-beta T cells. Subsets of gamma-delta T cells have shown antitumor and immunoregulatory activity.Specifications
TCR V beta 8b | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.5% BSA and 0.1% sodium azide | |
0 | |
TRB | |
Human TCR Vbeta8 | |
200 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, T-Cell Activation | |
MX-6 | |
FITC | |
TRB | |
T cell receptor beta locus; TCRB; TRB; TRB@ | |
Mouse | |
Protein G | |
RUO | |
6957 | |
4° C, store in dark | |
Liquid |