Learn More
Teknorinse™ Acidid Rinse Aid
Teknorinse enables water to drain evenly from washed items so that they are left virtually dry
Brand: Teknon TK310/5
Additional Details : Weight : 5.03000kg
Description
Teknorinse enables water to drain evenly from washed items so that they are left virtually dry. It also helps to neutralise the alkaline detergent used for the wash cycle. This formulation is designed to operate in hard or soft water and helps prevent scale and promotes efficiency. It is highly economical having an in-use concentration in the order of 300ppm.- Rinse neutraliser
- Scale control
- Low foam
- Free rinsing An acidic rinse aid for use in laboratory, medical and industrial automatic washing machines.
Specifications
Teknorinse Acidic Rinse Aid | |
For use in laboratory, medical and industrial automatic washing machines |