Learn More
BINDER™ Telescopic Rack
Product Code. 30238255 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB Series
KB/KB PRO 260, KB/KB PRO 470, KBF/KBF PRO 260
KBF PRO Series
KBF Series
Unit Size:
Each
Product Code. 30238255
Brand: BINDER™ 80122385
Only null left
Add to basket
Add to basket
Shelf rack in-set with pair of ball bearing mounted telescopic retaining rails.
  • Shelf rack made of stainless steel
  • Includes set of telescopic rails
Specifications
For Use With (Equipment) KBF PRO Series
Material Stainless Steel
Type Rack