Learn More
Memmert™ Temperature Restriction for UF1060plus
Product Code. 17538835 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF1060plus Oven
Unit Size:
Each
Product Code. 17538835
Brand: Memmert™ UF1060plusA8
Only null left
Add to basket
Add to basket
Optional temperature restriction between 60°C and 250°C.
  • Accessory
  • 1 Choice
  • Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
For Use With (Equipment) UF1060plus Oven
Type Temperature Restriction