Learn More
Eppendorf™ Temperature Sensor Device for Mastercycler
Product Code. p-7159263 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
Mastercycler, 384-well
Mastercycler, 96-well
Unit Size:
Each
Product Code. 15733145
Brand: Eppendorf™ 0056001000
Rapid, easy to use, and suitable for use with Mastercycler PCR cyclers.
  • Measuring device without temperature sensor
  • Sensor for 96 well and 384 well blocks, sold separately
  • Simple, rapid verification and adjustment
  • Automatic adjustment with Mastercycler software, version 2.10 or higher
  • Can be third-party adjusted
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Requires Temperature Sensor
For Use With (Equipment) Mastercycler, 96-well
Description Single Channel