Learn More
IKA™ Test Tube Attachment for IKA Platform Shaker
Attachment with customized array for holding reagent glasses on a platform shaker.
32.00 EUR - 65.00 EUR
Specifications
Product Type | Test Tube Adapter Attachment |
---|---|
Description | Test tube adapter attachment |
For Use With (Equipment) | Universal attachment, shaker |
Product Code | Brand | Holds | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Product Type | Price | Quantity & Availability | ||||
10490243
|
IKA™
L001830 |
Test Tube Adapter Attachment |
32.00 EUR
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
10728971
|
IKA™
L001840 |
Tubes | Test Tube Adapter Attachment |
65.00 EUR
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
||||
10130921
|
IKA™
L001850 |
Tubes | Test Tube Adapter Attachment |
65.00 EUR
Each |
Estimated Shipment: 25-11-2024
Log in to see stock. |
||||
Description
Attachment with customized array for holding reagent glasses on a platform shaker.
- Requires VG 3.3 universal attachment
- Facilitates multi-tube vortexing
Specifications
Test Tube Adapter Attachment | |
Test tube adapter attachment | |
Universal attachment, shaker |