Learn More
Memmert™ Test Tube Rack
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
WTB35 Water Baths
WTB50 Water Baths
Holds:
100 Test Tubes (4 Rows)
30 Test Tubes (2 Rows)
44 Test Tubes (2 Rows)
50 Test Tubes (2 Rows)
58 Test Tubes (2 Rows)
66 Test Tubes (2 Rows)
74 Test Tubes (2 Rows)
75 Test Tubes (3 Rows)
Diameter (Metric):
10 mm
12 mm
14.5 mm
18 mm
21 mm
32 mm
Unit Size:
Each
Description
- Water Bath Accessory
Specifications
Specifications
| For Use With (Equipment) | WTB50 Water Baths |
| Type | Test Tube Rack |
| Holds | 50 Test Tubes (2 Rows) |
| Material | Stainless Steel |
| Diameter (Metric) | 18 mm |