Learn More
Memmert™ Test Tube Racks with 48 Holes
Product Code. 17110365 Shop All Memmert Products
Click to view available options
Features:
Longitudinals for sizes 15 and 24
Transverses for sizes 15 and 24
For Use With (Equipment):
WTB11
WTB15
WTB24
WTB6
Unit Size:
Each
Product Code. 17110365
Brand: Memmert™ WTB24B48837
Currently Unavailable: We have no information when this product will be available.
View alternative products
Test tube rack with 48 holes and 4-row rack
  • Test tube rack
  • 48 holes 4-row rack
  • Diameter is 18 mm
Specifications
Description 48 holes, 4-row rack
For Use With (Equipment) WTB24
Type Test Tube Rack
Features Longitudinals for sizes 15 and 24
Diameter (Metric) 18 mm