missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophenolphthalein ethyl ester potassium salt

Product Code. 11343306
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11343306

Brand: Thermo Scientific Alfa Aesar L12674.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62637-91-6 | C22H13Br4KO4 | 700.06 g/mol

Tetrabromophenolphthalein ethyl ester potassium salt is used as reagent for detection of proteins and as indicator for ion-pair extraction titration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromophenolphthalein ethyl ester potassium salt is used as reagent for detection of proteins and as indicator for ion-pair extraction titration.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-91-6
Molecular Formula C22H13Br4KO4
Molecular Weight (g/mol) 700.06
MDL Number MFCD00011662
InChI Key WCIQBKUTYDIBJC-UHFFFAOYSA-M
Synonym potassium tetrabromophenolphthalein ethyl ester, tetrabromophenolphthalein ethyl ester potassium salt, potassium 2,6-dibromo-4-3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene 2-ethoxycarbonyl phenyl methyl phenolate, potassium 2,6-dibromo-4-3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene 2-ethoxycarbonyl phenyl methyl benzenolate, dj-2, benzoic acid, 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-, ethyl ester, potassium salt, benzoic acid, 2-3,5-dibromo-4-hydroxyphenyl 3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene methyl-, ethyl ester, potassium salt 1:1, tetrabromophenolphthalein ethyl ester potassium, 3',3,5',5-tetrabromophenolphthalein ethyl ester potassium salt
PubChem CID 23689366
IUPAC Name potassium;2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)-(2-ethoxycarbonylphenyl)methyl]phenolate
SMILES [K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1

Specifications

Melting Point ∽273°C (decomposition)
Quantity 1 g
Beilstein 3848263
Solubility Information Soluble in water.
Formula Weight 700.06
Chemical Name or Material Tetrabromophenolphthalein ethyl ester potassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.