missing translation for 'onlineSavingsMsg'
Learn More

TG 100801, MedChemExpress

Product Code. p-200097889
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463618

Brand: Medchem Express HY10186

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TG 100801 is a prodrug that generates TG 100572 by de-esterification in development to treat age-related macular degeneration. TG 100572 is a multi-targeted kinase inhibitor which inhibits receptor tyrosine kinases and Src kinases; has IC50s of 2, 7, 2, 16, 13, 5, 0.5, 6, 0.1, 0.4, 1, 0.2 for VEGFR1, VEGFR2, FGFR1, FGFR2, PDGFRβ, Fgr, Fyn, Hck, Lck, Lyn, Src, Yes, respectively.

TG 100801 is a prodrug that generates TG 100572 by de-esterification in development to treat age-related macular degeneration. TG 100572 is a multi-targeted kinase inhibitor which inhibits receptor tyrosine kinases and Src kinases

Specifications

Chemical Name or Material TG 100801
CAS 867331-82-6
Color Red
Purity Grade Notes Research
Molecular Formula C33H30ClN5O3
Quantity 10 mg
Solubility Information DMSO : 5.56 mg/mL (9.58 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C1)OC2=CC=C(Cl)C(C3=CC(C)=C(N=C(NC(C=C4)=CC=C4OCCN5CCCC5)N=N6)C6=C3)=C2
Molecular Weight (g/mol) 580.08
Formula Weight 580.08
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.