Learn More
Thermo Scientific™ Wash Buffer 1 for MagJET Plasmid DNA Kit (concentrated)
Wash lysed samples to remove any contaminants or inhibitors from the isolated plasmid DNA. Thermo Scientific™ MagJET™ Wash Buffer 1 (concentrated) is a component of the MagJET™ Plasmid DNA Kits (K2791/2792) and may be purchased separately in 110mL aliquots.
Brand: Thermo Scientific™ R2611
49.50 EUR valid until 2024-12-14
Use promo code "21632" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price
Description
The Thermo Scientific™ MagJET™ Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Wash Buffer | |
MagJET | |
110 mL |
For Research Use Only. Not for use in diagnostic procedures.