missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WELQut Protease (5 U/μL)

Thermo Scientific™ WELQut Protease is a highly specific serine protease cleaving outside the WELQX recognition motif to remove N-terminal protein fusion tags.

Brand:  Thermo Scientific™ EO0861

Product Code. 13243149

  • 161.00 EUR / 500 units
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ WELQut Protease is a highly specific serine protease cleaving outside the WELQX recognition motif to remove N-terminal protein fusion tags.

Thermo Scientific WELQut Protease is highly specific, recombinant serine protease of Staphylococcus aureus. It recognizes and precisely cleaves recombinant proteins containing an engineered recognition sequence† W- E- L- Q↓X (Trp, Glu, Leu, Gln, X can be any amino acid). The protease cleaves outside the recognition sequence without leaving additional amino acids bound to the target protein.
The WELQut Protease is active in a broad temperature (4 to 30°C) and pH (pH 6.5 to 9.0) range and does not require specific buffers.
In addition, this new protease has several procedural advantages - it is ideal for on-column proteolysis reactions and can be easily removed from reaction mixtures using its built-in His-tag.

Highlights

  • Cleaves outside WELQ recognition sequence, without leaving additional amino acids bound to the target protein
  • Highly specific to cognate recognition site, does not generate non-specific product bands, even after long incubation and using excess of protease
  • Easy to remove from the reaction mixture using built-in His-tag
  • Ideal for on-column proteolysis reactions
Applications
  • Removal of N-terminal fusion tags from recombinant protein preparations.
Footnote
† This cleavage sequence is present in expression vector pLATE52 included into the Thermo Scientific aLICator LIC Cloning and Expression Kit 4 (N-terminal His-tag/WELQut) (#K1281) available from Thermo Scientific.

Definition of Activity Unit: Each unit is defined as the amount of enzyme required to cleave ≥ 99% of 100 μg of a control protein in 16 h at 20°C.

  • Enzyme activity is assayed in 100 μL 100 mM Tris-HCl (pH 8.0)
  • Hazardous: No
  • Molecular Weight: 22 kDa monomer
  • Source:Bacillus subtilis cells with a cloned gene of SplB protease from Staphylococcus aureus
  • Storage Buffer: Enzyme is supplied in 10 mM Na2HPO4; 1.8 mM KH2PO4, pH 7.3; 140 mM NaCl; 2.7 mM KCl, 50% glycerol
  • Storage Condition:-20°C
  • -20°C

TRUSTED_SUSTAINABILITY
Specifications

Specifications

WELQut
500 units
Protease
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.