Learn More
Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple
Shop All Cole Parmer Products
Click to view available options
Length (English):
120 in. (probe)
60 in. (probe)
Unit Size:
Each
Pack of 6
Description
- Flexible Kapton-insulated wire is ideal for checking food temperatures
- Flexible fiberglass-Insulated wire used for high-temperature measurements
- Custom designs or changes to this product are available upon request
- Exposed-junction probes are not recommended for use with aqueous solutions
- Do not subject type J exposed-junction probes to moisture as they will rust
Specifications
Specifications
| Temperature (Metric) | -150°C to +482°C |
| Type | Thermocouple Probe |
| For Use With (Application) | Used for High-Temperature Measurements |
| Length (English) | 120 in. (probe) |