Learn More
Cole-Parmer™ Thermohygrometer
Shop All Cole Parmer Products
Click to view available options
Temperature Range (Metric):
-10°C to +50°C
Unit Size:
Each
Specifications
Specifications
| Type | Thermohygrometer |
| Accuracy | ± 2.5% |
| Temperature Range (Metric) | -10°C to +50°C |
| Material | Brass |
| Mounting | Brass |
| Diameter (Metric) Outer | 150 mm |
| Weight (Metric) | 320 g |
| Includes | White Face and Brass Case |