Learn More
Quickfit™ Thermoplastic Polyester Tubing Clamps
For use with flexible rubber and plastic tubing
Brand:  Quickfit KT6
31.26 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11332025
  • 32.90 EUR / Pack of 5
Estimated Shipment: 06-02-2025
Description
For use with flexible rubber and plastic tubing
  • Provides fine flow control of liquids and gases
  • Easy one handed adjustment: just roll serated wheel along sloped floor
  • Manufactured from thermoplastic polyester
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tubing Clamp
Up to 6mm O.D. tubing
5/Pk.
Thermoplastic Polyester
Used with flexible Rubber and Plastic tubing