missing translation for 'onlineSavingsMsg'
Learn More

Thiodiglycolic acid, 98%, Thermo Scientific Chemicals

Product Code. 11385139
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it applications in the production of [1,4]oxathiane-2,6-dione by heating. It is employed as an anti-oxidant and reducing agent in cosmetics industry.

Solubility
Solubility in water is almost transparent. (water, 4.00E+05 mg/L @ 25°C (exp); 1e+006 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-93-3
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.148
MDL Number MFCD00004359
InChI Key UVZICZIVKIMRNE-UHFFFAOYSA-N
Synonym thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
PubChem CID 31277
ChEBI CHEBI:35012
IUPAC Name 2-(carboxymethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCC(=O)O

Specifications

Melting Point 127°C to 131°C
UN Number UN3261
Beilstein 1764392
Merck Index 149331
Formula Weight 150.15
Percent Purity 98%
Quantity 100 g
Chemical Name or Material Thiodiglycolic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'