Learn More
Gravis Wide Neck Glass Bottles
Product Code. 16421394
Click to view available options
Capacity (Metric):
60 mL
Provides excellent storage for liquids and solids in the laboratory. Parisienne de Verrerie™ Wide Neck Glass Bottles are to be sealed with waterproof screw caps.
  • Wide neck allows for ease of access to contents
  • Manufactured from soda lime glass
Specifications
Capacity (Metric) 60 mL
Material Glass
No. per Case 72Pack
Shape Round