missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test
Rapidly detect fibrin and fibrinogen degradation products in human serum and urine by latex agglutination.
73.40 EUR - 477.00 EUR
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
13215669
|
Thermo Scientific™
R30852301 |
Thrombo-Wellcotest Latex | 3 mL/Vial | 2°C to 8°C |
107.00 EUR
3mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13255629
|
Thermo Scientific™
R30852601 |
Thrombo-Wellcotest Kit | 20 Tests/Kit | 2°C to 8°C | N/A | ||||
13295539
|
Thermo Scientific™
R30853001 |
Thrombo-Wellcotest Collection Tubes | 100 Tubes/Pk. | 2°C to 8°C |
477.00 EUR
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
13255509
|
Thermo Scientific™
R30853101 |
Thrombo-Wellcotest Glycine Saline Buffer | 100 mL/Bottle | 2°C to 8°C |
73.40 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Carry out semi-quantitative detection of fibrin and fibrinogen degradation products in human serum and urine using Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test.
Use this simple, rapid, semi-quantitative test for the presence of fibrin/fibrinogen degredation products (FDP) in human serum and urine samples.
The test is designed as a slide agglutination method in which one drop of sample and one drop of latex suspension are mixed. A distinct agglutination pattern at the end of the test period indicates the presence of 2μg/ml FDP or more in the sample under test. By assaying samples at two different dilutions, an approximate FDP value can be determined.
Capable of detecting the presence of all the major breakdown products of fibrin or fibrinogen. The levels measured by this latex test correlate well with results obtained with a haemagglutination-inhibition immunoassay.
The following products are available for use with Thrombo-Wellcotest Test Kit and are sold separately:
The test is designed as a slide agglutination method in which one drop of sample and one drop of latex suspension are mixed. A distinct agglutination pattern at the end of the test period indicates the presence of 2μg/ml FDP or more in the sample under test. By assaying samples at two different dilutions, an approximate FDP value can be determined.
Capable of detecting the presence of all the major breakdown products of fibrin or fibrinogen. The levels measured by this latex test correlate well with results obtained with a haemagglutination-inhibition immunoassay.
The following products are available for use with Thrombo-Wellcotest Test Kit and are sold separately:
Thrombo-Wellcotest Latex (Part No. R30852301)
Thrombo-Wellcotest Collection Tubes (Part No. R30853001)—Glass tubes contain soya bean trypsin inhibitor (approximately 3600 NF units/tube) and Bothrops atrox venom (>10μg/tube) for the collection of 2mL whole blood or urine.
Thrombo-Wellcotest Glycine Saline Buffer (Part No. R30853101)—Glycine saline buffer is provided as a replacement item in instances where multiple tests have exhausted the supply of this component.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title