missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIAR Mouse anti-Human, Unconjugated, Clone: 1E8D12, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | TIAR |
---|---|
Clone | 1E8D12 |
Concentration | 940 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829195
|
Proteintech
66907-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819195
|
Proteintech
66907-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TIAR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TIAR. This antibody recognizes human, mouse, rat antigen. The TIAR antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
TIAR | |
940 μg/mL | |
Monoclonal | |
Mouse | |
7073 | |
Primary |
1E8D12 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |