missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFAIP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23456-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody, which is a rabbit polyclonal antibody raised against 350 aa of C-terminus of human TNFAIP3, can recognize full-length A20 (∽90 kDa) and cleaved form (∽37-40 kDa).
This gene was identified as a gene whose expression is rapidly induced by the tumor necrosis factor (TNF). The protein encoded by this gene is a zinc finger protein, and has been shown to inhibit NF-kappa B activation as well as TNF-mediated apoptosis. Knockout studies of a similar gene in mice suggested that this gene is critical for limiting inflammation by terminating TNF-induced NF-kappa B responses.Specifications
TNFAIP3 | |
Polyclonal | |
Unconjugated | |
TNFAIP3 | |
A20, OTUD7C, TNF alpha induced protein 3, TNFA1P2, TNFAIP3, Zinc finger protein A20 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21929, 683206, 7128 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21580, Q60769 | |
TNFAIP3 | |
TNFAIP3 Fusion Protein Ag18150 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |