Learn More
Tocris Bioscience™ AC 187
Potent and selective amylin antagonist
Brand:  Tocris Bioscience™ 3419/500U
Product Code. 18034502
  • 437.00 EUR / 500µg
Description
AC 187 is an orally active, potent amylin receptor antagonist (IC50 = 0.48 nM) that displays 38-fold and 400-fold selectivity over calcitonin and CGRP receptors respectively. Blocks amyloid β-induced neurotoxicity by attenuating the activation of initiator and effector caspases in vitro. Increases glucagon secretion, accelerates gastric emptying, alters plasma glucose levels and increases food intake in vivo.
Specifications
Specifications
0.95
Store at -20°C
500 μg