Learn More
Tocris Bioscience™ [Nle4,D-Phe7]-α-MSH
Melanocortin receptor agonist
Brand:  Tocris Bioscience™ 3013/1
202.74 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18055272
  • 259.00 EUR / 1mg
Explore more special offers
Description
Description
[Nle4,D-Phe7]-α-MSH is a synthetic analog of α-MSH that is an agonist at melanocortin receptors (Ki values are 0.085, 0.4, 3.8 and 5.1 nM for MC1, MC3, MC4 and MC5 receptors respectively).