missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene-d8, for NMR, 100.0 atom % D, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for NMR
Specifications
Specifications
Physical Form | Liquid |
Color | Colorless |
Density | 0.94g/mL |
Deuterium (D) | 99.95% min. |
Quantity | 5 g |
CAS | 2037-26-5 |
Molecular Formula | C7H8 |
Linear Formula | C6D5CD3 |
Synonym | toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d |
InChI Key | YXFVVABEGXRONW-JGUCLWPXSA-N |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'