missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Extra pure, Honeywell Burdick & Jackson™
Click to view available options
173 kg
Specifications
Specifications
Melting Point | -95°C |
CAS | 108-88-3 |
Color | Colorless |
CAS Max % | 1 |
Density | 0.867 g/cm3 (20°C) |
Boiling Point | 110.6°C |
Quantity | 173 kg |
Flash Point | 4°C |
Molecular Formula | C7H8 |
Beilstein | 635760 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'