missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM22 Mouse anti-Human, Clone: 1C2B11, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | TOMM22 |
---|---|
Clone | 1C2B11 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828555
|
Proteintech
66562-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16818555
|
Proteintech
66562-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
66562-1-Ig recognizes a22 kDa protein that is larger than the calculated MW of 15 kDa.
Tom22 has a negatively charged N-terminal region exposed to the cytosol, a putative transmembrane region, and a C-terminal intermembrane space region with little negative charge. The following functions can be assigned to the three domains of Tom22: the cytosolic N-terminal domain plays a dual role, specifically important for presequence binding; the intermembrane space domain provides a trans binding site for presequences, and the single membrane anchor of Tom22 is crucial for the integrity of the GIP (general import gene) complex. The TOM complex of mammalian mitochondria resembles the fungal Tom complex, but is distinct from the plant TOM system. Thus, while unique components of the mammalian mitochondrial import system have been identified (e. g. TOM34 and metaxin), Tom22, and Tom37 have not been identified in plant mitochondria.Specifications
TOMM22 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
1C9 2, hTom22, MST065, MSTP065, TOM22, TOMM22 | |
TOMM22 | |
IgG2a | |
Protein A | |
Antibody |
1C2B11 | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9NS69 | |
56993 | |
TOMM22 Fusion Protein Ag1797 | |
Primary | |
-20°C | |
TOMM22 |