Learn More
Fisherbrand™ Top-Line Pipette Tips
Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.5 to 20 μL
1 to 200 μL
5 to 300 μL
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Length (Metric):
34.1mm
45.7mm
50.8mm
55mm
Unit Size:
Pack of 1000
Pack of 960
Description
- Optimized cone geometry for a secure fit
- Graduation marks on tip at standard volumes
- Conical tip opening prevents drop formation at the end of the tip
- Tips made of autoclavable, non-wettable, virgin polypropylene
- Sterile packs available (beta irradiation)
Specifications
Specifications
| Volume (Metric) | 0.5 to 20 μL |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Length (Metric) | 45.7mm |
| Certifications/Compliance | ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600 |
| Sterilization Method | Beta radiation |
| Format | Rack |
| Tip Style | Universal |
| For Use With (Equipment) | Universal Fit |
| Show More |