Learn More
Amarell™ Total Immersion Low Temperature Laboratory Thermometer
Precision thermometer, low temperatures, enclosed scale, capillary lens front, total immersion, Richter-top finish with button, suitable for government verification
Brand:  Amarell™ L33008
 View more versions of this product
Product Code. 10529291
  • 184.00 EUR / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Description
  • Low temperature precision thermometer
  • Total immersion
  • Richter-top finish with button
  • Suitable for government verification
Specifications
Specifications
Toluene
Government testing
350 mm
30°C
-100°C to +30°C
1°C
DIN 12778
Total Immersion
Low Temperature Thermometer
-100°C
Yes