Learn More
Amarell™ Total Immersion Low Temperature Laboratory Thermometer
Precision thermometer, low temperatures, enclosed scale, capillary lens front, total immersion, Richter-top finish with button, suitable for government verification
174.00 EUR - 184.00 EUR
Product Code | Brand | Temperature Range (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10733523
|
Amarell™
L33040 |
-100°C to +30°C | 400 mm | N/A | |||||
10529291
|
Amarell™
L33008 |
-100°C to +30°C | 350 mm |
184.00 EUR
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
||||
Description
- Low temperature precision thermometer
- Total immersion
- Richter-top finish with button
- Suitable for government verification
Specifications
Total Immersion |