Learn More
Fisherbrand™ TraceableOne™ Single-Use USB Temperature Data Logger
Shop All Cole Parmer Products
Click to view available options
Temperature Range (Metric):
-20°C to -15°C
2°C to 8°C
Data Memory:
16,000 Readings, 10 Day, 1 min. Interval
16,000 Readings, 30 Day, 3 min. Interval
16,000 Readings, 60 Day, 6 min. Interval
16,000 Readings, 90 Day, 10 min. Interval
Unit Size:
Pack of 40
Description
- LED alarm indicator for visual status indication
- Time delay of 30 minutes before logging begins for enhanced logging accuracy
- Protective plastic pouch maintains waterproof IP67 rating
- Bar code for in-house scanning
- Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Specifications
| Data Memory | 16,000 Readings, 30 Day, 3 min. Interval |
| Temperature Range (Metric) | -20°C to -15°C |