missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56178SS
This item is not returnable.
View return policy
Description
TRAF-5 Polyclonal specifically detects TRAF-5 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| TRAF-5 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| MGC:39780, RNF84RING finger protein 84, TNF receptor-associated factor 5 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| TRAF5 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| TRAF5 | |
| A synthetic peptide corresponding to amino acids 1-20 ([BLAST]MAHSEEQAAVPCAFIRQNSG[/BLAST]) of mouse TRAF5 was used as immunogen, GenBank no. AAH12702.1 | |
| 0.025 mL | |
| Apoptosis, Signal Transduction, Zinc Finger | |
| 7188 | |
| Human, Mouse, Rat, Gerbil | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction