missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
213.00 EUR - 453.00 EUR
Specifications
| Antigen | TRAF-6 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 3-5 ug/ml, Flow Cytometry |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066354
|
Novus Biologicals
NB100-56436SS |
0.025 mg |
213.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18765393
|
Novus Biologicals
NB100-56436 |
453.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TRAF-6 Polyclonal specifically detects TRAF-6 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| TRAF-6 | |
| Western Blot 3-5 ug/ml, Flow Cytometry | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Q9Y4K3 | |
| 7189 | |
| Anti-TRAF6 polyclonal antibody was raised against a peptide corresponding to amino acids between 410 and 460 of human TRAF6. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Signal Transduction, Zinc Finger | |
| 0.1 ml PBS with 0.02% Sodium Azide | |
| E3 ubiquitin-protein ligase TRAF6, EC 6.3.2.-, Interleukin-1 signal transducer, RING finger protein 85, RNF85MGC:3310, TNF receptor-associated factor 6 | |
| TRAF6 | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title