missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody (12A668) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 11 publications
234.61 EUR - 514.03 EUR
Specifications
Antigen | TRANCE/TNFSF11/RANK L |
---|---|
Clone | 12A668 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 25859979), Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1 - 5 ug/ml, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready |
Applications | Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18082426
|
Bio-Techne
NB100-56593SS |
0.025 mg |
234.61 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755703
|
Novus Biologicals
NB100-56593 |
0.1 mg |
514.03 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
TRANCE/TNFSF11/RANK L | |
1.0 mg/mL | |
Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Cancer | |
O14788 | |
8600 | |
A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen (NP_003692). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
12A668 | |
Western Blot 1-2 ug/ml, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 25859979), Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1 - 5 ug/ml, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
TNFSF11 | |
IgG1 κ | |
Protein G purified | |
35 kDa |
For Research Use Only