missing translation for 'onlineSavingsMsg'
Learn More

trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane, 97%, Thermo Scientific Chemicals

Product Code. 15477006
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15477006

Brand: Thermo Scientific Alfa Aesar H62087.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917342-29-1
Molecular Formula C13H25NO3
Molecular Weight (g/mol) 243.347
MDL Number MFCD03844626
InChI Key KTNFSGIXLVLQNK-UHFFFAOYSA-N
Synonym tert-butyl trans-4-2-hydroxyethyl-cyclohexylcarbamate, tert-butyl cis-4-2-hydroxyethyl-cyclohexylcarbamate, tert-butyl trans-4-2-hydroxyethyl cyclohexyl carbamate, tert-butyl trans-4-2-hydroxyethyl cyclohexylcarbamate, tert-butyl cis-4-2-hydroxyethyl cyclohexylcarbamate, tert-butyl n-1r,4r-4-2-hydroxyethyl cyclohexyl carbamate, ktnfsgixlvlqnk-xypyzodxsa-n, butyl trans-4-2-hydroxyethyl cyclohexyl carbamate, tert-butyl n-4-2-hydroxyethyl cyclohexyl carbamate
PubChem CID 24720929
IUPAC Name tert-butyl N-[4-(2-hydroxyethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CCO

Specifications

Quantity 250 mg
Formula Weight 243.34
Percent Purity 97%
Chemical Name or Material trans-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.