missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Benzylamino)cyclohexanol, 99%, Thermo Scientific™

Product Code. 11469380
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40571-86-6
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD00145421
InChI Key NJNCYFUGUYIMEQ-CHWSQXEVSA-N
Synonym 1r,2r-2-benzylamino cyclohexanol, trans-2-benzylamino cyclohexanol, trans-2-benzylamino-1-cyclohexanol, 1r,2r-2-benzylamino cyclohexan-1-ol, 1r,2r-2-benzylamino-1-cyclohexanol, cyclohexanol, 2-phenylmethyl amino-, 1r-trans, 2-benzylamino-cyclohexanol, trans-2-benzylamino-cyclohexanol, pubchem19547, trans-2-benzylaminocyclohexan-1-ol
PubChem CID 2724650
IUPAC Name (1R,2R)-2-(benzylamino)cyclohexan-1-ol
SMILES C1CCC(C(C1)NCC2=CC=CC=C2)O

Specifications

Melting Point 67°C to 70°C
Quantity 5 g
Formula Weight 205.3
Percent Purity 99%
Chemical Name or Material trans-2-(Benzylamino)cyclohexanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.