Learn More
Fisherbrand™ Transparent Terylene Mesh PVC Tubing
Product Code. 12345869 Shop All Fisherbrand Products
Click to view available options
Description:
6.5 x 3mm
5 x 3mm
8 x 3mm
10 x 3mm
Unit Size:
10m
Product Code. 12345869
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
General purpose tubing for transfer of fluids, gases, and powdered foods
All dimensions are approximate.
  • Transparent, colorless
  • Terylene mesh reinforced wall
  • Flexible yet great inherent strength
  • Suitable for moderate pressure or vacuum applications
Specifications
Description 6.5 x 3mm
Material PVC
Overall Length (Metric) 10 m
For Use With (Application) Suitable For Moderate Pressure Or Vacuum Applications
Packaging Quantity 1 Pack
Color Transparent