missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trastuzumab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Trastuzumab |
---|---|
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Hybridoma |
Regulatory Status | RUO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478119
|
ACROBiosystems
TRB-Y1-100ug |
100 μg |
POA
100µg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16488119
|
ACROBiosystems
TRB-Y1-1mg |
1 mg |
POA
1mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Trastuzumab Monoclonal antibody specifically detects Trastuzumab.Specifications
Trastuzumab | |
Unconjugated | |
RUO | |
-20°C |
Monoclonal | |
Hybridoma | |
Primary |