Learn More
uvex Trend Black/Blue PU 69358 Boot - Width 11
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Brand:  uvex 6935844
94.63 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17243731
  • 107.00 EUR / 1 pair
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Description
Uvex 2 Trend Boot 69358 S2 SRC
  • Sporty S2 safety boot with wide fit
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • Smooth-running, dirt and heat-resistant lacing system
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole
    • Moisture transport system helps maintain foot temperature
  • Soft padding collar and dust tongue
  • Removable and anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 69358
  • Standard EN ISO 20345:2011 S2 SRC
  • Sizes: 38 to 52
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black, Blue
44
Steel
EN ISO 20345:2011
Standard Width
Yes
S2
ESD Safety Shoe
Leather
Dual-density polyurethane (PU/PU)
Reusable
Unisex
Uvex 2
No
SRC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
uvex Trend Black/Blue PU 69358 Boot - Width 11 > Black/Blue, 69358 Boot, Width: 11, Size: 44