missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid Solution, MP Biomedicals™

Artikelnummer. 11480605
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Trichloroacetic acid Solution

  • It is used in protein precipitation; has been used to determine protein concentration by quantitative precipitation.
  • It is also Used as a decalcifier and fixative in microscopy.
  • A protocol for the precipitation of nucleic acids can be found in Molecular Cloning. Rats exposed to chronic sublethal amounts of trichloroacetic acid displayed an increase in serum bilirubin with a decrease in hematological proteins and cholesterols along with significant decreases in red blood corpuscles, mean cell volume, mean corpuscular hemoglobin, hemoglobin and hematocrit.
  • In mice the production of liver tumors was associated to Trichloroacetic acid, which is also a known mouse hepatocarinogen.
  • On the guinea pig trichloroacetic acid was observed to be a mild alergen on the skin.
  • Trichloroacetic acid is an acetic acid analogue commonly used to precipitate proteins, DNA and RNA. In the presence of SDS Trichloroacetic acid will precipitate proteins which can then be quantified by the Lowry method.
  • The compound has also found uses as a decalcifier and fixative in microscopy, in protein sequencing, detecting albumin and organic synthesis.
TRUSTED_SUSTAINABILITY

Spezifikation

pH 1.2 (0.1 M aqueous solution)(literature)
CAS 76-03-9
Melting Point ∼56°C (literature)
Boiling Point 197°C at 1013 hPa(Literature)
Molecular Formula C2HCl3O2
MDL Number MFCD00004177
UN Number UN2564
Beilstein 970119
Synonym Trichloroethanoic Acid, TCA, Aceto-caustin, Acide trichloracetique
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Molecular Weight (g/mol) 163.38
Density 1.61 g/cm3 (Lit.)
Quantity 500 mL
Physical Form Solution
Vapor Density 5.6 (air = 1)(Lit.)
Vapor Pressure 1.2 hPa at 50°C(Lit.)
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'