Learn More
Tris(dioxa-3,6-heptyl)amine, 95%, Thermo Scientific Chemicals
59.60 EUR - 627.00 EUR
Chemical Identifiers
CAS | 70384-51-9 |
---|---|
Molecular Formula | C15H33NO6 |
Molecular Weight (g/mol) | 323.42 |
MDL Number | MFCD00010748 |
InChI Key | XGLVDUUYFKXKPL-UHFFFAOYSA-N |
Synonym | tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine |
PubChem CID | 112414 |
IUPAC Name | 2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine |
SMILES | COCCOCCN(CCOCCOC)CCOCCOC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10512521
|
Thermo Scientific Acros
124850250 |
25 mL | Glass bottle |
59.60 EUR
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10753691
|
Thermo Scientific Acros
124851000 |
100 mL | Glass bottle |
151.00 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10465675
|
Thermo Scientific Acros
124855000 |
500 mL | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
70384-51-9 | |
323.42 | |
XGLVDUUYFKXKPL-UHFFFAOYSA-N | |
112414 | |
COCCOCCN(CCOCCOC)CCOCCOC |
C15H33NO6 | |
MFCD00010748 | |
tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine | |
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine |
Specifications
70384-51-9 | |
1.0100g/mL | |
162°C | |
95% | |
(CH3OCH2CH2OCH2CH2)3N | |
MFCD00010748 | |
1.01 | |
Solubility in water: miscible. Other solubilities: soluble in acetone, ether, toluene and cyclohexane | |
COCCOCCN(CCOCCOC)CCOCCOC | |
323.42 | |
323.42 | |
Liquid |
Yellow to Brown | |
330.0°C | |
Authentic | |
C15H33NO6 | |
1.4466 to 1.4486 | |
15,356; 16,373 | |
tris 2-2-methoxyethoxy ethyl amine, tris dioxa-3,6-heptyl amine, tris 3,6-dioxaheptyl amine, tda-1, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl ethanamine, ethanamine, 2-2-methoxyethoxy-n,n-bis 2-2-methoxyethoxy ethyl, 8-2-2-methoxyethoxy ethyl-2,5,11,14-tetraoxa-8-azapentadecane, tris-2-2-methoxyethoxy ethyl amine | |
XGLVDUUYFKXKPL-UHFFFAOYSA-N | |
2-(2-methoxyethoxy)-N,N-bis[2-(2-methoxyethoxy)ethyl]ethanamine | |
112414 | |
95% | |
Tris(dioxa-3, 6-heptyl)amine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 274-590-5
RUO – Research Use Only